missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Activin A / INHBA Protein

Recombinant Protein

Brand:  enQuireBio™ QP5357-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955969

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_002183.1
Activin A / INHBA Protein
Research Use Only
INHBA
Recombinant Protein
E. coli
MGLECDGKVN ICCKKQFFVS FKDIGWNDWI IAPSGYHANY CEGECPSHIA GTSGSSLSFH STVINHYRMR GHSPFANLKS CCVPTKLRPM SMLYYDDGQN IIKKDIQNMI VEECGCS
Greater than 95% as determined by SDS-PAGE.
3624
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 10 ng/mL in the Cytotoxicity of MPC-11 cells assay. The bioactivity acceptance criteria is set at 1 x 10^5 units/mg.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers