missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACY3 Protein
A cDNA sequence encoding the ACY3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10958-1mg
Specifications
ACY3 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ACY3 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 0.1M NaCl. |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMCSLPVP REPLRRVAVT GGTHGNEMSG VYLARHWLHA PAELQRASFS AVPVLANPAA TSGCRRYVDHDLNRTFTSSF LNSRPTPDDP YEVTRARELN QLLGPKASGQ AFDFVLDLHN TTANMGTCLI AKSSHEVFAM HLCRHLQLQY PELSCQVFLY QRSGEESYNL DSVAKNGLGL ELGPQPQGVL RADIFSRMRT LVATVLDFIE LFNQGTAFPA FEMEAYRPVG VVDFPRTEAG HLAGTVHPQL QDRDFQPLQP GAPIFQMFSG EDLLYEGEST VYPVFINEAA YYEKGVAFVQ TEKFTFTVPA MPALTPAPSP AS |