missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADH6 Protein
A cDNA sequence encoding the ADH6 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10967-5ug
Additional Details : Weight : 0.01000kg
Specifications
130 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSTTGQ VIRCKAAILW KPGAPFSIEE VEVAPPKAKE VRIKVVATGL CGTEMKVLGS KHLDLLYPTI LGHEGAGIVE SIGEGVSTVK PGDKVITLFL PQCGECTSCL NSEGNFCIQF KQSKTQLMSD GTSRFTCKGK SIYHFGNTST FCEYTVIKEI SVAKIDAVAP LEKVCLISCG FSTGFGAAIN TAKVTPGSTC AVFGLGGVGL SVVMGCKAAG AARIIGVDVN KEKFKKAQEL GATECLNPQD LKKPIQEVLF DMTDAGIDFC FEAIGNLDVL AAALASCNES YGVCVVVGVL PASVQLKISG QLFFSGRSLK GSVFGGWKSR QHIPKLVADY MAEKLNLDPL ITHTLNLDKI NEAVELMKTG KCIRCILLL | |
Greater than 90% as determined by SDS-PAGE. |
ADH6 Protein | |
Research Use Only | |
ADH6 | |
Human | |
His | |
The ADH6 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 30% glycerol, 0.15M NaCl and 1mM DTT. |