missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Adiponectin / Acrp30 / ADIPOQ Protein
Recombinant Protein
107.00 € - 4060.00 €
Specifications
Accession Number | NP_001171271.1 |
---|---|
Gene ID (Entrez) | 9370 |
Name | Adiponectin / Acrp30 / ADIPOQ Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 5.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15995829
|
enQuireBio™
QP5279-5UG |
5 μg |
107.00 €
5µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975829
|
enQuireBio™
QP5279-25UG |
25 μg |
252.00 €
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955829
|
enQuireBio™
QP5279-100UG |
100 μg |
630.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985829
|
enQuireBio™
QP5279-500UG |
500 μg |
2490.00 €
500µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965829
|
enQuireBio™
QP5279-1MG |
1 mg |
4060.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001171271.1 | |
Adiponectin / Acrp30 / ADIPOQ Protein | |
Less than or equal to 5.00 EUs/ug as determined by the LAL method. | |
gACRP (globular adiponectin) activity was confirmed by measuring inhibition of cytotoxicity of M1 cells. Typical ED50 values for our recombinant adiponectin in this assay are in the range of 3 ug/ml. | |
Human | |
Untagged | |
10 mM Tris, 0.75 mM DTT, pH 8.0 |
9370 | |
Research Use Only | |
ADIPOQ | |
Recombinant Protein | |
E. coli | |
MKGEPGEGAY VYRSAFSVGL ETYVTIPNMP IRFTKIFYNQ QNHYDGSTGK FHCNIPGLYY FAYHITVYMK DVKVSLFKKD KAMLFTYDQY QENNVDQASG SVLLHLEVGD QVWLQVYGEG ERNGLYADND NDSTFTGFLL YHDTN | |
Greater than 95% as determined by SDS-PAGE. |