missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADPRH Protein

A cDNA sequence encoding the ADPRH was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10971-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985728

  • 3401.86 € / 1mg

Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

141
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ADPRH
Human
His
The ADPRH solution (0.5 mg/ml) contains 20mM Tris-HCl buffer, pH 8.0, 10% glycerol, 1mM DTT and 100mM NaCl.
ADPRH Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMEKYVA AMVLSAAGDA LGYYNGKWEF LQDGEKIHRQ LAQLGGLDAL DVGRWRVSDD TVMHLATAEA LVEAGKAPKL TQLYYLLAKH YQDCMEDMDG RAPGGASVHN AMQLKPGKPN GWRIPFNSHE GGCGAAMRAM CIGLRFPHHS QLDTLIQVSI ESGRMTHHHP TGYLGALASA LFTAYAVNSR PPLQWGKGLM ELLPEAKKYI VQSGYFVEEN LQHWSYFQTK WENYLKLRGI LDGESAPTFP ESFGVKERDQ FYTSLSYSGW GGSSGHDAPM IAYDAVLAAG DSWKELAHRA FFHGGDSDST AAIAGCWWGV MYGFKGVSPS NYEKLEYRNRLEETARALYS LGSKEDTVIS L
Videos
SDS
Documents
Certificates