missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AGA / ASRG / Aspartylglucosaminidase Protein
A DNA sequence encoding the Homo sapiens (Human) AGA / ASRG / Aspartylglucosaminidase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5620-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000018.2 | |
AGA / ASRG / Aspartylglucosaminidase Protein | |
Research Use Only | |
AGA | |
Recombinant Protein | |
E. coli | |
SSPLPLVVNT WPFKNATEAA WRALASGGSA LDAVESGCAM CEREQCDGSV GFGGSPDELG ETTLDAMIMD GTTMDVGAVG DLRRIKNAIG VARKVLEHTT HTLLVGESAT TFAQSMGFIN EDLSTTASQA LHSDWLARNC QPNYWRNVIP DPSKYCGPYK PPGILKQDIP IHKETEDDRG HD | |
Greater than 90% as determined by SDS-PAGE. |
175 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |