missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AGR3 Protein

A cDNA sequence encoding the AGR3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10980-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985758

  • 3401.86 € / 1mg

Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

AGR3 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
AGR3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMIAIKKE KRPPQTLSRG WGDDITWVQT YEEGLFYAQK SKKPLMVIHH LEDCQYSQAL KKVFAQNEEI QEMAQNKFIM LNLMHETTDK NLSPDGQYVP RIMFVDPSLT VRADIAGRYS NRLYTYEPRD LPLLIENMKK ALRLIQSEL
Videos
SDS
Documents
Certificates