missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AITR Protein

A cDNA sequence encoding the AITR was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10989-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985788

  • 203.00 € / 10µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

8784
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Baculovirus
QRPTGGPGCG PGRLLLGTGT DARCCRVHTT RCCRDYPGEE CCSEWDCMCV QPEFHCGDPC CTTCRHHPCP PGQGVQSQGK FSFGFQCIDC ASGTFSGGHE GHCKPWTDCT QFGFLTVFPG NKTHNAVCVP GSPPAEPLEH HHHHH
Greater than 95.0% as determined by SDS-PAGE.
AITR Protein
Research Use Only
TNFRSF18
Human
His
AITR protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers