missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human alpha-Galactosidase A / GLA Protein
A DNA sequence encoding the Homo sapiens (Human) alpha-Galactosidase A / GLA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6086-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000160.1 | |
alpha-Galactosidase A / GLA Protein | |
Research Use Only | |
GLA | |
Recombinant Protein | |
E. coli | |
LDNGLARTPT MGWLHWERFM CNLDCQEEPD SCISEKLFME MAELMVSEGW KDAGYEYLCI DDCWMAPQRD SEGRLQADPQ RFPHGIRQLA NYVHSKGLKL GIYADVGNKT CAGFPGSFGY YDIDAQTFAD WGVDLLKFDG CYCDSLENLA DGYKHMSLAL NRTGRSIVYS CEWPLYMWPF QKPNYTEIRQ YCNHWRNFAD IDDSWKSIKS ILDWTSFNQE RIVDVAGPGG WNDPDMLVIG NFGLSWNQQV TQMALWAIMA APLFMSNDLR HISPQAKALL QDKDVIAINQ DPLGKQGYQL RQGDNFEVWE RPLSGLAWAV AMINRQEIGG PRSYTIAVAS LGKGVACNPA CFITQLLPVK RKLGFYEWTS RLRSHINPTG TVLLQLENTM QMSLKDLL | |
Greater than 90% as determined by SDS-PAGE. |
2717 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |