missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Annexin A3 Protein
A DNA sequence encoding the Homo sapiens (Human) Annexin A3, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_005130.1 |
---|---|
Gene ID (Entrez) | 306 |
Name | Annexin A3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16017011
|
enQuireBio™
QP8890-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037011
|
enQuireBio™
QP8890-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027011
|
enQuireBio™
QP8890-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047011
|
enQuireBio™
QP8890-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057011
|
enQuireBio™
QP8890-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067011
|
enQuireBio™
QP8890-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_005130.1 | |
Annexin A3 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
306 | |
Research Use Only | |
ANXA3 | |
Recombinant Protein | |
E. coli | |
WVGHRGTVRD YPDFSPSVDA EAIQKAIRGI GTDEKMLISI LTERSNAQRQ LIVKEYQAAY GKELKDDLKG DLSGHFEHLM VALVTPPAVF DAKQLKKSMK GAGTNEDALI EILTTRTSRQ MKDISQAYYT VYKKSLGDDI SSETSGDFRK ALLTLADGRR DESLKVDEHL AKQDAQILYK AGENRWGTDE DKFTEILCLR SFPQLKLTFD EYRNISQKDI VDSIKGELSG HFEDLLLAIV NCVRNTPAFL AERLHRALKG IGTDEFTLNR IMVSRSEIDL LDIRTEFKKH YGYSLYSAIK SDTSGDYEIT LLKICGGD | |
Greater than 90% as determined by SDS-PAGE. |