missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Annexin A8-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Annexin A8-like protein 1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001092315.2 |
---|---|
Gene ID (Entrez) | 728113 |
Name | Annexin A8-like protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16020341
|
enQuireBio™
QP7711-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16040341
|
enQuireBio™
QP7711-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16030341
|
enQuireBio™
QP7711-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050341
|
enQuireBio™
QP7711-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060341
|
enQuireBio™
QP7711-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070341
|
enQuireBio™
QP7711-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001092315.2 | |
Annexin A8-like protein 1 | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
728113 | |
Research Use Only | |
ANXA8L2 | |
Recombinant Protein | |
E. coli | |
MAWWKAWIEQ EGVTVKSSSH FNPDPDAETL YKAMKGIGVG SQLLSHQAAA FAFPSSALTS VSPWGQQGHL CCNPAGTNEQ AIIDVLTKRS NTQRQQIAKS FKAQFGKDLT ETLKSELSGK FERLIVALMY PPYRYEAKEL HDAMKGSRDD VSSFVDPALA LQDAQDLYAA GEKIRGTDEM KFITILCTRS ATHLLRVKCT QNLHSYFAER LYYAMKGAGT RDGTLIRNIV SRSEIDLNLI KCHFKKMYGK TLSSMIMEDT SGDYKNALLS LVGSDP | |
Greater than 80% as determined by SDS-PAGE. |