missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ANXA7 Protein
A cDNA sequence encoding the ANXA7 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | ANXA7 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15975938
|
enQuireBio™
QP11036-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965938
|
enQuireBio™
QP11036-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955938
|
enQuireBio™
QP11036-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ANXA7 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ANXA7 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 150mM NaCl, 1mM DTT and 40% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSYPGY PPTGYPPFPG YPPAGQESSF PPSGQYPYPS GFPPMGGGAY PQVPSSGYPG AGGYPAPGGY PAPGGYPGAP QPGGAPSYPG VPPGQGFGVP PGGAGFSGYP QPPSQSYGGG PAQVPLPGGF PGGQMPSQYP GGQPTYPSQP ATVTQVTQGT IRPAANFDAI RDAEILRKAM KGFGTDEQAI VDVVANRSND QRQKIKAAFK TSYGKDLIKD LKSELSGNME ELILALFMPP TYYDAWSLRK AMQGAGTQER VLIEILCTRT NQEIREIVRC YQSEFGRDLE KDIRSDTSGH FERLLVSMCQ GNRDENQSIN HQMAQEDAQR LYQAGEGRLG TDESCFNMIL ATRSFPQLRA TMEAYSRMAN RDLLSSVSRE FSGYVESGLK TILQCALNRP AFFAERLYYA MKGAGTDDST LVRIVVTRSE IDLVQIKQMF AQMYQKTLGT MIAGDTSGDY RRLLLAIVGQ | |
Greater than 85.0% as determined by SDS-PAGE. |