missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APOA1 Protein
A cDNA sequence encoding the APOA1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11043-1mg
Additional Details : Weight : 0.01000kg
Specifications
335 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human | |
MKAAVLTLAV LFLTGSQARH FWQQDEPPQS PWDRVKDLAT VYVDVLKDSG RDYVSQFEGS ALGKQLNLKL LDNWDSVTST FSKLREQLGP VTQEFWDNLE KETEGLRQEM SKDLEEVKAK VQPYLDDFQK KWQEEMELYR QKVEPLRAEL QEGARQKLHE LQEKLSPLGE EMRDRARAHV DALRTHLAPY SDELRQRLAA RLEALKENGG ARLAEYHAKA TEHLSTLSEK AKPALEDLRQ GLLPVLESFK VSFLSALEEY TKKLNTQ | |
Greater than 95.0% as determined by SDS-PAGE. |
APOA1 Protein | |
Research Use Only | |
APOA1 | |
Human | |
Untagged | |
The APOA1 1 mg/ml solution contains 10mM Ammonium Bicarbonate at pH 7.4. |