missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APOH Protein
A cDNA sequence encoding the APOH was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Gene ID (Entrez) | 350 |
---|---|
Name | APOH Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | APOH |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15905998
|
enQuireBio™
QP11052-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985988
|
enQuireBio™
QP11052-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995988
|
enQuireBio™
QP11052-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
350 | |
Research Use Only | |
APOH | |
Human | |
His | |
APOH protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2M urea and 10% glycerol. |
APOH Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGRTCPKP DDLPFSTVVP LKTFYEPGEE ITYSCKPGYV SRGGMRKFIC PLTGLWPINT LKCTPRVCPF AGILENGAVR YTTFEYPNTI SFSCNTGFYL NGADSAKCTE EGKWSPELPV CAPIICPPPS IPTFATLRVY KPSAGNNSLY RDTAVFECLP QHAMFGNDTI TCTTHGNWTK LPECREVKCP FPSRPDNGFV NYPAKPTLYY KDKATFGCHD GYSLDGPEEI ECTKLGNWSA MPSCKASCKV PVKKATVVYQ GERVKIQEKF KNGMLHGDKV SFFCKNKEKK CSYTEDAQCI DGTIEVPKCF KEHSSLAFWK TDASDVKPC | |
Greater than 85.0% as determined by SDS-PAGE. |