missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human APOH Protein

A cDNA sequence encoding the APOH was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11052-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985988

Quantity Price
1 171.64 € / 10µg
10 161.27 € / 10µg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

350
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
APOH
Human
His
APOH protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2M urea and 10% glycerol.
APOH Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSGRTCPKP DDLPFSTVVP LKTFYEPGEE ITYSCKPGYV SRGGMRKFIC PLTGLWPINT LKCTPRVCPF AGILENGAVR YTTFEYPNTI SFSCNTGFYL NGADSAKCTE EGKWSPELPV CAPIICPPPS IPTFATLRVY KPSAGNNSLY RDTAVFECLP QHAMFGNDTI TCTTHGNWTK LPECREVKCP FPSRPDNGFV NYPAKPTLYY KDKATFGCHD GYSLDGPEEI ECTKLGNWSA MPSCKASCKV PVKKATVVYQ GERVKIQEKF KNGMLHGDKV SFFCKNKEKK CSYTEDAQCI DGTIEVPKCF KEHSSLAFWK TDASDVKPC
Videos
SDS
Documents
Certificates