missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Apolipoprotein C-IV Protein
A DNA sequence encoding the Homo sapiens (Human) Apolipoprotein C-IV, was expressed in the hosts and tags indicated.
287.00 € - 3175.00 €
Specifications
Accession Number | NP_001637.1 |
---|---|
Gene ID (Entrez) | 346 |
Name | Apolipoprotein C-IV Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16097411
|
enQuireBio™
QP8992-YE-100UG |
100 μg |
880.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037421
|
enQuireBio™
QP8992-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007421
|
enQuireBio™
QP8992-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027421
|
enQuireBio™
QP8992-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047421
|
enQuireBio™
QP8992-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017421
|
enQuireBio™
QP8992-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001637.1 | |
Apolipoprotein C-IV Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
346 | |
Research Use Only | |
APOC4 | |
Recombinant Protein | |
Yeast | |
CQPEAQEGTL SPPPKLKMSR WSLVRGRMKE LLETVVNRTR DGWQWFWSPS TFRGFMQTYY DDHLRDLGPL TKAWFLESKD SLLKKTHSLC PRLVCGDKDQ G | |
Greater than 90% as determined by SDS-PAGE. |