missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Apoptosis regulatory protein Siva Protein
A DNA sequence encoding the Homo sapiens (Human) Apoptosis regulatory protein Siva, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_006418.2 |
---|---|
Gene ID (Entrez) | 10572 |
Name | Apoptosis regulatory protein Siva Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16095120
|
enQuireBio™
QP6688-YE-100UG |
100 μg |
615.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16035130
|
enQuireBio™
QP6688-YE-500UG |
500 μg |
1665.00 €
500µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16005130
|
enQuireBio™
QP6688-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16025130
|
enQuireBio™
QP6688-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045130
|
enQuireBio™
QP6688-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015130
|
enQuireBio™
QP6688-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_006418.2 | |
Apoptosis regulatory protein Siva Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
10572 | |
Research Use Only | |
SIVA1 | |
Recombinant Protein | |
Yeast | |
MPKRSCPFAD VAPLQLKVRV SQRELSRGVC AERYSQEVFD PSGVASIACS SCVRAVDGKA VCGQCERALC GQCVRTCWGC GSVACTLCGL VDCSDMYEKV LCTSCAMFET | |
Greater than 90% as determined by SDS-PAGE. |