missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APRIL Protein
A cDNA sequence encoding the APRIL was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11057-5ug
Additional Details : Weight : 0.01000kg
Specifications
8741 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MASMTGGQQM GRGSHMAVLT QKQKKQHSVL HLVPINATSK DDSDVTEVMW QPALRRGRGL QAQGYGVRIQ DAGVYLLYSQ VLFQDVTFTM GQVVSREGQG RQETLFRCIR SMPSHPDRAY NSCYSAGVFH LHQGDILSVI IPRARAKLNL SPHGTFLGL | |
Greater than 85.0% as determined by SDS-PAGE. |
APRIL Protein | |
Research Use Only | |
TNFSF13 | |
Human | |
Untagged | |
APRIL protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |