missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Aquaporin-4 Protein
A DNA sequence encoding the Homo sapiens (Human) Aquaporin-4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5672-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001641.1 | |
Aquaporin-4 Protein | |
Research Use Only | |
AQP4 | |
Recombinant Protein | |
E. coli | |
CPDVEFKRRF KEAFSKAAQQ TKGSYMEVED NRSQVETDDL ILKPGVVHVI DVDRGEEKKG KDQSGEVLSS V | |
Greater than 81% as determined by SDS-PAGE. |
361 | |
1 mg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |