missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ARL5A Protein
A cDNA sequence encoding the ARL5A was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | ARL5A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15966058
|
enQuireBio™
QP11073-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946058
|
enQuireBio™
QP11073-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956058
|
enQuireBio™
QP11073-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ARL5A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ARL5A protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 0.2mM PMSF and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMGILFT RIWRLFNHQE HKVIIVGLDN AGKTTILYQF SMNEVVHTSP TIGSNVEEIV INNTRFLMWD IGGQESLRSS WNTYYTNTEF VIVVVDSTDR ERISVTREEL YKMLAHEDLR KAGLLIFANK QDVKECMTVA EISQFLKLTS IKDHQWHIQA CCALTGEGLC QGLEWMMSRL KIR | |
Greater than 90% as determined by SDS-PAGE. |