missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ASF1B Protein
A cDNA sequence encoding the ASF1B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11084-20ug
Additional Details : Weight : 0.01000kg
Specifications
ASF1B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAKVSVLNVA VLENPSPFHS PFRFEISFEC SEALADDLEW KIIYVGSAES EEFDQILDSV LVGPVPAGRH MFVFQADAPN PSLIPETDAV GVTVVLITCT YHGQEFIRVG YYVNNEYLNP ELRENPPMKP DFSQLQRNIL ASNPRVTRFH INWDNNMDRL EAIETQDPSL GCGLPLNCTPIKGLGLPGCI PGLLPENSMD CILEHHHHHH | |
Greater than 85% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ASF1B protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.15M NaCl and 1mM DTT. |