missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Aspartyl aminopeptidase Protein
A DNA sequence encoding the Homo sapiens (Human) Aspartyl aminopeptidase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5940-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001306049.1 | |
Aspartyl aminopeptidase Protein | |
Research Use Only | |
DNPEP | |
Recombinant Protein | |
E. coli | |
MQVAMNGKAR KEAVQTAAKE LLKFVNRSPS PFHAVAECRN RLLQAGFSEL KETEKWNIKP ESKYFMTRNS STIIAFAVGG QYVPGNGFSL IGAHTDSPCL RVKRRSRRSQ VGFQQVGVET YGGGIWSTWF DRDLTLAGRV IVKCPTSGRL EQQLVHVERP ILRIPHLAIH LQRNINENFG PNTEMHLVPI LATAIQEELE KGTPEPGPLN AVDERHHSVL MSLLCAHLGL SPKDIVEMEL CLADTQPAVL GGAYDEFIFA PRLDNLHSCF CALQALIDSC AGPGSLATEP HVRMVTLYDN EEVGSESAQG AQSLLTELVL RRISASCQHP TAFEEAIPKS FMISADMAHA VHPNYLDKHE ENHRPLFHKG PVIKVNSKQR YASNAVSEAL IREVANKVKV PLQDLMVRND TPCGTTIGPI LASRLGLRVL DLGSPQLAMH SIREMACTTG VLQTLTLFKG FFELFPSLSH NLLVD | |
Greater than 90% as determined by SDS-PAGE. |
23549 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |