missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Asporin Protein
A DNA sequence encoding the Homo sapiens (Human) Asporin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5687-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001180264.1 | |
Asporin Protein | |
Research Use Only | |
ASPN | |
Recombinant Protein | |
E. coli | |
DMEDTDDDDD DDDDDDDDDE DNSLFPTREP RSHFFPFDLF PMCPFGCQCY SRVVHCSDLG LTSVPTNIPF DTRMLDLQNN KIKEIKENDF KGLTSLYGLI LNNNKLTKIH PKAFLTTKKL RRLYLSHNQL SEIPLNLPKS LAELRIHENK VKKIQKDTFK GMNALHVLEM SANPLDNNGI EPGAFEGVTV FHIRIAEAKL TSVPKGLPPT LLELHLDYNK ISTVELEDFK RYKELQRLGL GNNKITDIEN GSLANIPRVR EIHLENNKLK KIPSGLPELK YLQIIFLHSN SIARVGVNDF CPTVPKMKKS LYSAISLFNN PVKYWEMQPA TFRCVLSRMS VQLGNFGM | |
Greater than 90% as determined by SDS-PAGE. |
54829 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |