missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Atrial natriuretic peptide receptor 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Atrial natriuretic peptide receptor 2, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_003986.2 |
---|---|
Gene ID (Entrez) | 4882 |
Name | Atrial natriuretic peptide receptor 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16033270
|
enQuireBio™
QP6432-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053270
|
enQuireBio™
QP6432-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043270
|
enQuireBio™
QP6432-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16063270
|
enQuireBio™
QP6432-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073270
|
enQuireBio™
QP6432-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083270
|
enQuireBio™
QP6432-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_003986.2 | |
Atrial natriuretic peptide receptor 2 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
4882 | |
Research Use Only | |
NPR2 | |
Recombinant Protein | |
E. coli | |
RNLTLAVVLP EHNLSYAWAW PRVGPAVALA VEALGRALPV DLRFVSSELE GACSEYLAPL SAVDLKLYHD PDLLLGPGCV YPAASVARFA SHWRLPLLTA GAVASGFSAK NDHYRTLVRT GPSAPKLGEF VVTLHGHFNW TARAALLYLD ARTDDRPHYF TIEGVFEALQ GSNLSVQHQV YAREPGGPEQ ATHFIRANGR IVYICGPLEM LHEILLQAQR ENLTNGDYVF FYLDVFGESL RAGPTRATGR PWQDNRTREQ AQALREAFQT VLVITYREPP NPEYQEFQNR LLIRAREDFG VELGPSLMNL IAGCFYDGIL LYAEVLNETI QEGGTREDGL RIVEKMQGRR YHGVTGLVVM DKNNDRETDF VLWAMGDLDS GDFQPAAHYS GAEKQIWWTG RPIPWVKGAP PSDNPPCAFD LDDPSCDKTP LSTLAI | |
Greater than 90% as determined by SDS-PAGE. |