missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BATF3 Protein
A cDNA sequence encoding the BATF3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11129-10ug
Additional Details : Weight : 0.01000kg
Specifications
55509 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSQGLPA AGSVLQRSVA APGNQPQPQP QQQSPEDDDR KVRRREKNRV AAQRSRKKQT QKADKLHEEY ESLEQENTML RREIGKLTEE LKHLTEALKE HEKMCPLLLC PMNFVPVPPR PDPVAGCLPR | |
Greater than 90.0% as determined by SDS-PAGE. |
BATF3 Protein | |
Research Use Only | |
BATF3 | |
Human | |
His | |
BATF3 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl and 30% glycerol. |