missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BCAT2 Protein
A DNA sequence encoding the Homo sapiens (Human) BCAT2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5707-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001158245.1 | |
BCAT2 Protein | |
Research Use Only | |
BCAT2 | |
Recombinant Protein | |
E. coli | |
ASSSFKAADL QLEMTQKPHK KPGPGEPLVF GKTFTDHMLM VEWNDKGWGQ PRIQPFQNLT LHPASSSLHY SLQLFEGMKA FKGKDQQVRL FRPWLNMDRM LRSAMRLCLP SFDKLELLEC IRRLIEVDKD WVPDAAGTSL YVRPVLIGNE PSLGVSQPTR ALLFVILCPV GAYFPGGSVT PVSLLADPAF IRAWVGGVGN YKLGGNYGPT VLVQQEALKR GCEQVLWLYG PDHQLTEVGT MNIFVYWTHE DGVLELVTPP LNGVILPGVV RQSLLDMAQT WGEFRVVERT ITMKQLLRAL EEGRVREVFG SGTACQVCPV HRILYKDRNL HIPTMENGPE LILRFQKELK EIQYGIRAHE WMFPV | |
Greater than 90% as determined by SDS-PAGE. |
587 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |