missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bcl 2 (minus NWGR domain) Protein
A cDNA sequence encoding the Bcl 2 (minus NWGR domain) was constructed and used to recombinantly synthesize the protein.
131.00 € - 6960.00 €
Specifications
Name | Bcl 2 (minus NWGR domain) Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15994038
|
enQuireBio™
QP10494-HIS-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974038
|
enQuireBio™
QP10494-HIS-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984038
|
enQuireBio™
QP10494-HIS-1MG |
1 mg |
6960.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Bcl 2 (minus NWGR domain) Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The protein contains 10mM Tris-HCL pH 8.0, 1mM EDTA and 250mM NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAHAGRTGYD NREIVMKYIH YKLSQRGYEW DAGDVGAAPP GAAPAPGIFS SQPGHTPHPA ASRDPVARTS PLQTPAAPGA AAGPALSPVP PVVHLTLRQA GDDFSRRYRR DFAEMSSQLH LTPFTARGRF ATVVEELFRD GVNWGRIVAF FEFGGVMCVE SVNREMSPLV DNIALWMTEY LNRHLHTWIQ DNGGWDAFVE LYGPSMRPLF DFSWLSLKTL LSLALVGACI TLGAYLGHK | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |