missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Bcl 6 Protein

A cDNA sequence encoding the Bcl 6 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11139-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15986268

Quantity Price
1 228.51 € / 2µg
6 214.69 € / 2µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

604
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MASPADSCIQ FTRHASDVLL NLNRLRSRDI LTDVVIVVSR EQFRAHKTVL MACSGLFYSI FTDQLKCNLS VINLDPEINP EGFCILLDFM YTSRLNLREG NIMAVMATAM YLQMEHVVDT CRKFIKASEA EMVSAIKPPR EEFLNSRMLM PQDIMAYRGR EVVENNLPLR SAPGCESRAF APSLYSGLST PPASYSMYSH LPVSSLLFSD EEFRDVRMPV ANPFPKERAL PCDSARPVPG EYSRPTLEVS PNVCHSNIYS PKETIPEEAR SDMHYSVAEG LKPAAPSARN APYFPCDKAS KEEERPSSED EIALHFEPPN APLNRKGLVS PQSPQKSDCQ PNSPTESCSS KNACILQASG SPPAKSPTDP KACNWKKYKF IVLNSLNQNA KPEGPEQAEL GRLSPRAYTA PPACQPPMEP ENLDLQSPTK LSASGEDSTI PQASRLNNIV NRSMTGSPRS SSESHSPLYM HPPKCTSCGS QSPQHAEMCL HTAGPTFPEE MGETQSEYSD SSCENGAFFC NECDCRFSEE ASLKRHTLQT HSDKPYKCDR CQASFRYKGN LASHKTVHTG EKPYRCNICG AQFNRPANLK THTRIHSGEK PYKCETCGAR FVQVAHLRAH VLIHTGEKPY PCEICGTRFR HLQTLKSHLR IHTGEKPYHC EKCNLHFRHK SQLRLHLRQK HGAITNTKVQ YRVSATDLPP ELPKAC
Bcl 6 Protein
Research Use Only
BCL6
Human
GST
Bcl-6 protein in 50mM Tris-HCl, pH 7.5 and 10mM L-glutathione (reduced).
Videos
SDS
Documents
Certificates