missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BCL2 Protein
A cDNA sequence encoding the BCL2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11142-20ug
Additional Details : Weight : 0.01000kg
Specifications
BCL2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
AHAGRTGYDN REIVMKYIHY KLSQRGYEWD AGDVGAAPPG AAPAPGIFSS QPGHTPHPAA SRDPVARTSP LQTPAAPGAA AGPALSPVPP VVHLTLRQAG DDFSRRYRRD FAEMSSQLHL TPFTARGRFA TVVEELFRDG VNWGRIVAFF EFGGVMCVES VNREMSPLVD NIALWMTEYL NRHLHTWIQD NGGWDAFVEL YGPSMRPLFD | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
BCL2 is a 0.2microm filtered concentrated solution which contains 25 mM Tris-HCl, pH 8.0, 100mM NaCl, 1 mM DTT, 30 % Glycerol, and 0.01%Tween-80. |