missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BD5 Protein
A cDNA sequence encoding the BD5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11149-1mg
Additional Details : Weight : 0.01000kg
Specifications
504180; 245908 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
GLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR I | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
BD5 Protein | |
Research Use Only | |
DEFB105A; DEFB105B | |
Human | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |