missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BEN domain-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) BEN domain-containing protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7716-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16040371

  • 2285.00 € / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001073919.1
BEN domain-containing protein 1
Research Use Only
BEND3
Recombinant Protein
E. coli
CLPQLNDFFS RFWAQREMED SQPSGQVASF FEAEQVDPGH FLDNKDQEEA LSLDRSSTIA SDHVVDTQDL TEFLDEASSP GEFAVFLLHR LFPELFDHRK LGEQYSCYGD GGKQELDPQR LQIIRNYTEI YFPD
Greater than 90% as determined by SDS-PAGE.
57673
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers