missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Beta-1,4-galactosyltransferase 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Beta-1,4-galactosyltransferase 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8332-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001186802.1 | |
Beta-1,4-galactosyltransferase 3 Protein | |
Research Use Only | |
B4GALT3 | |
Recombinant Protein | |
E. coli | |
LSALFGRDQG PTFDYSHPRD VYSNLSHLPG APGGPPAPQG LPYCPERSPL LVGPVSVSFS PVPSLAEIVE RNPRVEPGGR YRPAGCEPRS RTAIIVPHRA REHHLRLLLY HLHPFLQRQQ LAYGIYVIHQ AGNGTFNRAK LLNVGVREAL RDEEWDCLFL HDVDLLPEND HNLYVCDPRG PRHVAVAMNK FGYSLPYPQY FGGVSALTPD QYLKMNGFPN EYWGWGGEDD DIATRVRLAG MKISRPPTSV GHYKMVKHRG DKGNEENPHR FDLLVRTQNS WTQDGMNSLT YQLLARELGP LYTNITADIG TDPRGPRAPS GPRYPPGSSQ AFRQEMLQRR PPARPGPLST ANHTALRGSH | |
Greater than 90% as determined by SDS-PAGE. |
8703 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |