missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Biliverdin reductase A Protein
A DNA sequence encoding the Homo sapiens (Human) Biliverdin reductase A, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_000703.2 |
---|---|
Gene ID (Entrez) | 644 |
Name | Biliverdin reductase A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15987259
|
enQuireBio™
QP5721-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15907269
|
enQuireBio™
QP5721-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15997259
|
enQuireBio™
QP5721-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15917269
|
enQuireBio™
QP5721-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15927269
|
enQuireBio™
QP5721-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15937269
|
enQuireBio™
QP5721-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000703.2 | |
Biliverdin reductase A Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
644 | |
Research Use Only | |
BLVRA | |
Recombinant Protein | |
E. coli | |
AEPERKFGVV VVGVGRAGSV RMRDLRNPHP SSAFLNLIGF VSRRELGSID GVQQISLEDA LSSQEVEVAY ICSESSSHED YIRQFLNAGK HVLVEYPMTL SLAAAQELWE LAEQKGKVLH EEHVELLMEE FAFLKKEVVG KDLLKGSLLF TAGPLEEERF GFPAFSGISR LTWLVSLFGE LSLVSATLEE RKEDQYMKMT VCLETEKKSP LSWIEEKGPG LKRNRYLSFH FKSGSLENVP NVGVNKNIFL KDQNIFVQKL LGQFSEKELA AEKKRILHCL GLAEEIQKYC CSRK | |
Greater than 80% as determined by SDS-PAGE. |