missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Biliverdin reductase A Protein

A DNA sequence encoding the Homo sapiens (Human) Biliverdin reductase A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5721-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987259

Quantity Price
1 434.28 € / 100µg
6 408.22 € / 100µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

NP_000703.2
Biliverdin reductase A Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
644
Greater than 80% as determined by SDS-PAGE.
Research Use Only
BLVRA
Recombinant Protein
E. coli
AEPERKFGVV VVGVGRAGSV RMRDLRNPHP SSAFLNLIGF VSRRELGSID GVQQISLEDA LSSQEVEVAY ICSESSSHED YIRQFLNAGK HVLVEYPMTL SLAAAQELWE LAEQKGKVLH EEHVELLMEE FAFLKKEVVG KDLLKGSLLF TAGPLEEERF GFPAFSGISR LTWLVSLFGE LSLVSATLEE RKEDQYMKMT VCLETEKKSP LSWIEEKGPG LKRNRYLSFH FKSGSLENVP NVGVNKNIFL KDQNIFVQKL LGQFSEKELA AEKKRILHCL GLAEEIQKYC CSRK
Videos
SDS
Documents
Certificates