Learn More
enQuireBio™ Recombinant Human BLyS / TNFSF13B / BAFF Protein
Recombinant Protein
Brand:  enQuireBio™ QP5331-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15985919
  • 6169.88 € / 1mg
Estimated Shipment: 26-11-2021
Specifications
NP_001139117.1
BLyS / TNFSF13B / BAFF Protein
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MHHHHHHLVP RAVQGPEETV TQDCLQLIAD SETPTIQKGS YTFVPWLLSF KRGSALEEKE NKILVKETGY FFIYGQVLYT DKTYAMGHLI QRKKVHVFGD ELSLVTLFRC IQNMPETLPN NSCYSAGIAK LEEGDELQLA IPRENAQISL DGDVTFFGAL KLL
10673
Greater than 90 as determined by SDS-PAGE.
Research Use Only
TNFSF13B
Human
Untagged
10 mM sodium phosphate, pH 7.5