missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BLyS / TNFSF13B / BAFF Protein

A DNA sequence encoding the Homo sapiens (Human) BLyS / TNFSF13B / BAFF, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8298-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16033861

  • 505.00 € / 100µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001139117.1
BLyS / TNFSF13B / BAFF Protein
Research Use Only
TNFSF13B
Recombinant Protein
E. coli
QVAALQGDLA SLRAELQGHH AEKLPAGAGA PKAGLEEAPA VTAGLKIFEP PAPGEGNSSQ NSRNKRAVQG PEETVTQDCL QLIADSETPT IQKGSYTFVP WLLSFKRGSA LEEKENKILV KETGYFFIYG QVLYTDKTYA MGHLIQRKKV HVFGDELSLV TLFRCIQNMP ETLPNNSCYS AGIAKLEEGD ELQLAIPREN AQISLDGDVT FFGALKLL
Greater than 90% as determined by SDS-PAGE.
10673
100 μg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers