missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BOLA3 Protein

A cDNA sequence encoding the BOLA3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11175-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15986378

Quantity Price
1 114.77 € / 5µg
10 107.33 € / 5µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

BOLA3 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
BOLA3 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAAWSPA AAAPLLRGIR GLPLHHRMFA TQTEGELRVT QILKEKFPRA TAIKVTDISG GCGAMYEIKI ESEEFKEKRT VQQHQMVNQA LKEEIKEMHG LRIFTSVPKR
Videos
SDS
Documents
Certificates