missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BP-1 Protein
A cDNA sequence encoding the BP-1 was constructed and used to recombinantly synthesize the protein.
264.00 € - 650.00 €
Specifications
Gene ID (Entrez) | 1748 |
---|---|
Name | BP-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | DLX4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15966448
|
enQuireBio™
QP11197-2UG |
2 μg |
264.00 €
2µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976448
|
enQuireBio™
QP11197-5UG |
5 μg |
397.00 €
5µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956448
|
enQuireBio™
QP11197-10UG |
10 μg |
650.00 €
10µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
1748 | |
Research Use Only | |
DLX4 | |
Human | |
Untagged | |
BP-1 protein in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |
BP-1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MTSLPCPLPG RDASKAVFPD LAPVPSVAAA YPLGLSPTTA ASPNLSYSRP YGHLLSYPYT EPANPGDSYL SCQQPAALSQ PLCGPAEHPQ ELEADSEKPR LSPEPSERRP QAPAKKLRKP RTIYSSLQLQ HLNQRFQHTQ YLALPERAQL AAQLGLTQTQ VKIWFQNKRS KYKKLLKQNS GGQEGDFPGR TFSVSPCSPP LPSLWDLPKA GTLPTSGYGN SFGAWYQHHS SDVLASPQMM |