missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BPI Protein
A cDNA sequence encoding the BPI was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | BPI Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15906458
|
enQuireBio™
QP11198-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986448
|
enQuireBio™
QP11198-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996448
|
enQuireBio™
QP11198-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
BPI Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
BPI (1.15 mg/ml) is supplied in 25mM Sodium Acetate pH 4.6. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRENMARGPC NAPRWASLMV LVAIGTAVTA AVNPGVVVRI SQKGLDYASQ QGTAALQKEL KRIKIPDYSD SFKIKHLGKG HYSFYSMDIR EFQLPSSQIS MVPNVGLKFS ISNANIKISG KWKAQKRFLK MSGNFDLSIE GMSISADLKL GSNPTSGKPT ITCSSCSSHI NSVHVHISKS KVGWLIQLFH KKIESALRNK MNSQVCEKVT NSVSSELQPY FQTLPVMTKI DSVAGINYGL VAPPATTAET LDVQMKGEFY SENHHNPPPF APPVMEFPAA HDRMVYLGLS DYFFNTAGLV YQEAGVLKMT LRDDMIPKES KFRLTTKFFG TFLPEVAKKF PNMKIQIHVS ASTPPHLSVQ PTGLTFYPAV DVQAFAVLPN SSLASLFLIG MHTTGSMEVS AESNRLVGEL KLDRLLLELK HSNIGPFPVE LLQDIMNYIV PILVLPRVNE KLQKGFPLPT PARVQLYNVV LQPHQNFLLF GADVVYK | |
Greater than 70% as determined by SDS-PAGE and capillary electrophoresis. |