missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BPI Protein
A cDNA sequence encoding the BPI was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11198-10ug
Additional Details : Weight : 0.01000kg
Specifications
BPI Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRENMARGPC NAPRWASLMV LVAIGTAVTA AVNPGVVVRI SQKGLDYASQ QGTAALQKEL KRIKIPDYSD SFKIKHLGKG HYSFYSMDIR EFQLPSSQIS MVPNVGLKFS ISNANIKISG KWKAQKRFLK MSGNFDLSIE GMSISADLKL GSNPTSGKPT ITCSSCSSHI NSVHVHISKS KVGWLIQLFH KKIESALRNK MNSQVCEKVT NSVSSELQPY FQTLPVMTKI DSVAGINYGL VAPPATTAET LDVQMKGEFY SENHHNPPPF APPVMEFPAA HDRMVYLGLS DYFFNTAGLV YQEAGVLKMT LRDDMIPKES KFRLTTKFFG TFLPEVAKKF PNMKIQIHVS ASTPPHLSVQ PTGLTFYPAV DVQAFAVLPN SSLASLFLIG MHTTGSMEVS AESNRLVGEL KLDRLLLELK HSNIGPFPVE LLQDIMNYIV PILVLPRVNE KLQKGFPLPT PARVQLYNVV LQPHQNFLLF GADVVYK | |
Greater than 70% as determined by SDS-PAGE and capillary electrophoresis. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
BPI (1.15 mg/ml) is supplied in 25mM Sodium Acetate pH 4.6. |