missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Breast carcinoma-amplified sequence 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Breast carcinoma-amplified sequence 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5705-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987149

Quantity Price
1 1443.46 € / 500µg
3 1356.85 € / 500µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

NP_003648.2
Breast carcinoma-amplified sequence 1 Protein
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
8537
Greater than 90% as determined by SDS-PAGE.
Research Use Only
BCAS1
Recombinant Protein
Yeast
MGNQMSVPQR VEDQENEPEA ETYQDNASAL NGVPVVVSTH TVQHLEEVDL GISVKTDNVA TSSPETTEIS AVADANGKNL GKEAKPEAPA AKSRFFLMLS RPVPGRTGDQ AADSSLGSVK LDVSSNKAPA NKDPSESWTL PVAAGPGQDT DKTPGHAPAQ DKVLSAARDP TLLPPETGGA GGEAPSKPKD SSFFDKFFKL DKGQEKVPGD SQQEAKRAEH QDKVDEVPGL SGQSDDVPAG KDIVDGKEKE GQELGTADCS VPGDPEGLET AKDDSQAAAI AENNNSIMSF FKTLVSPNKA ETKKDPEDTG AEKSPTTSAD LKSDKANFTS QETQGAGKNS KGCNPSGHTQ SVTTPEPAKE GTKEKSGPTS LPLGKLFWKK SVKEDSVPTG AEENVVCESP VEIIKSKEVE SALQTVDLNE GDAAPEPTEA KLKREESKPR TSLMAFLRQM SVKGDGGITH SEEINGKDSS CQTSDSTEKT ITPPEPEPTG APQKGKEGSS KDKKSAAEMN KQKSNKQEAK EPAQCTEQAT VDTNSLQNGD KLQKRPEKRQ QSLGGFFKGL GPKRMLDAQV QTDPVSIGPV GKSK
Videos
SDS
Documents
Certificates