missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C14ORF129 Protein

A cDNA sequence encoding the C14ORF129 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11222-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15986528

Quantity Price
1 114.77 € / 5µg
10 107.33 € / 5µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

C14ORF129 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The C14ORF129 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 50mM NaCl.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH METDCNPMEL SSMSGFEEGS ELNGFEGTDM KDMRLEAEAV VNDVLFAVNN MFVSKSLRCA DDVAYINVET KERNRYCLEL TEAGLKVVGY AFDQVDDHLQ TPYHETVYSL LDTLSPAYRE AFGNALLQRL EALKRDGQS
Videos
SDS
Documents
Certificates