missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C1D Protein
A DNA sequence encoding the Homo sapiens (Human) C1D, was expressed in the hosts and tags indicated.
228.00 € - 2440.00 €
Specifications
Accession Number | NP_001177192.1 |
---|---|
Gene ID (Entrez) | 10438 |
Name | C1D Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16098201
|
enQuireBio™
QP938-10UG |
10 μg |
228.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038211
|
enQuireBio™
QP938-50UG |
50 μg |
324.00 €
50µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088201
|
enQuireBio™
QP938-100UG |
100 μg |
555.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018211
|
enQuireBio™
QP938-200UG |
200 μg |
930.00 €
200µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028211
|
enQuireBio™
QP938-500UG |
500 μg |
1535.00 €
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16008211
|
enQuireBio™
QP938-1MG |
1 mg |
2440.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001177192.1 | |
C1D Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
10438 | |
Research Use Only | |
C1D | |
Recombinant Protein | |
E. coli | |
MAGEEINEDY PVEIHEYLSA FENSIGAVDE MLKTMMSVSR NELLQKLDPL EQAKVDLVSA YTLNSMFWVY LATQGVNPKE HPVKQELERI RVYMNRVKEI TDKKKAGKLD RGAASRFVKN ALWEPKSKNA SKVAN | |
Greater than 90% as determined by SDS-PAGE. |