missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C1QTNF4 Protein
A cDNA sequence encoding the C1QTNF4 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | C1QTNF4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15986558
|
enQuireBio™
QP11232-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966558
|
enQuireBio™
QP11232-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976558
|
enQuireBio™
QP11232-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
C1QTNF4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
C1QTNF4 filtered (0.4microm) solution at a concentration of 0.25 mg/ml in phosphate buffered saline. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
LGPTPGPGSS ELRSAFSAAR TTPLEGTSEM AVTFDKVYVN IGGDFDVATG QFRCRVPGAY FFSFTAGKAP HKSLSVMLVR NRDEVQALAF DEQRRPGARR AASQSAMLQL DYGDTVWLRL HGAPQYALGA PGATFSGYLV YADADADAPA RGPPAPPEPR SAFSAARTRS LVGSDAGPGP RHQPLAFDTE FVNIGGDFDA AAGVFRCRLP GAYFFSFTLG KLPRKTLSVK LMKNRDEVQA MIYDDGASRR REMQSQSVML ALRRGDAVWL LSHDHDGYGA YSNHGKYITF SGFLVYPDLA PAAPPGLGAS ELL HHHHHH | |
Greater than 85.0% as determined by SDS-PAGE. |