missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C6ORF108 Protein
A cDNA sequence encoding the C6ORF108 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | C6ORF108 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15956588
|
enQuireBio™
QP11241-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946588
|
enQuireBio™
QP11241-25UG |
25 μg |
203.00 €
25µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936588
|
enQuireBio™
QP11241-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
C6ORF108 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The C6ORF108 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8), 1mM DTT, and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAAAMVPGRS ESWERGEPGR PALYFCGSIR GGREDRTLYE RIVSRLRRFG TVLTEHVAAA ELGARGEEAAGGDRLIHEQD LEWLQQADVV VAEVTQPSLG VGYELGRAVA FNKRILCLFR PQSGRVLSAM IRGAADGSRF QVWDYEEGEV EALLDRYFEA DPPGQVAASP DPTT | |
Greater than 95.0% as determined by SDS-PAGE. |