missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C9ORF95 Protein
A cDNA sequence encoding the C9ORF95 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11245-1mg
Additional Details : Weight : 0.01000kg
Specifications
C9ORF95 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
C9ORF95 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer, pH 8.0, 10% glycerol, 2mM DTT and 200mM NaCl. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMKTFIIG ISGVTNSGKT TLAKNLQKHL PNCSVISQDD FFKPESEIET DKNGFLQYDV LEALNMEKMM SAISCWMESA RHSVVSTDQE SAEEIPILII EGFLLFNYKP LDTIWNRSYF LTIPYEECKR RRSTRVYQPP DSPGYFDGHV WPMYLKYRQE MQDITWEVVY LDGTKSEEDL FLQVYEDLIQ ELAKQKCLQV TA |