missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CA11 Protein
A cDNA sequence encoding the CA11 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 770 |
---|---|
Name | CA11 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CA11 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15906608
|
enQuireBio™
QP11246-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996598
|
enQuireBio™
QP11246-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986598
|
enQuireBio™
QP11246-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
770 | |
Research Use Only | |
CA11 | |
Human | |
His | |
The CA11 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
CA11 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MHIGPAPDPE DWWSYKDNLQ GNFVPGPPFW GLVNAAWSLC AVGKRQSPVD VELKRVLYDP FLPPLRLSTG GEKLRGTLYN TGRHVSFLPA PRPVVNVSGG PLLYSHRLSE LRLLFGARDG AGSEHQINHQ GFSAEVQLIH FNQELYGNFS AASRGPNGLA ILSLFVNVAS TSNPFLSRLL NRDTITRISY KNDAYFLQDL SLELLFPESF GFITYQGSLS TPPCSETVTW ILIDRALNIT SLQMHSLRLL SQNPPSQIFQ SLSGNSRPLQ PLAHRALRGN RDPRHPERRC RGPNYRLHVD GVPHGR | |
Greater than 85% as determined by SDS-PAGE. |