missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calcineurin subunit B type 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Calcineurin subunit B type 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8006-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_671709.1 | |
Calcineurin subunit B type 2 Protein | |
Research Use Only | |
PPP3R2 | |
Recombinant Protein | |
E. coli | |
GNEASYPAEM CSHFDNDEIK RLGRRFKKLD LDKSGSLSVE EFMSLPELRH NPLVRRVIDV FDTDGDGEVD FKEFILGTSQ FSVKGDEEQK LRFAFSIYDM DKDGYISNGE LFQVLKMMVG NNLTDWQLQQ LVDKTIIILD KDGDGKISFE EFSAVVRDLE IHKKLVLIV | |
Greater than 80% as determined by SDS-PAGE. |
5535 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |