missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Calponin-3 Protein

A DNA sequence encoding the Homo sapiens (Human) Calponin-3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5849-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988209

Quantity Price
1 1979.08 € / 1mg
3 1861.92 € / 1mg
Estimated Shipment: 24-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

NP_001272984.1
Calponin-3 Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
1266
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CNN3
Recombinant Protein
E. coli
THFNKGPSYG LSAEVKNKIA SKYDHQAEED LRNWIEEVTG MSIGPNFQLG LKDGIILCEL INKLQPGSVK KVNESSLNWP QLENIGNFIK AIQAYGMKPH DIFEANDLFE NGNMTQVQTT LVALAGLAKT KGFHTTIDIG VKYAEKQTRR FDEGKLKAGQ SVIGLQMGTN KCASQAGMTA YGTRRHLYDP KMQTDKPFDQ TTISLQMGTN KGASQAGMLA PGTRRDIYDQ KLTLQPVDNS TISLQMGTNK VASQKGMSVY GLGRQVYDPK YCAAPTEPVI HNGSQGTGTN GSEISDSDYQ AEYPDEYHGE YQDDYPRDYQ YSDQGIDY
Videos
SDS
Documents
Certificates