missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CARHSP1 Protein
A cDNA sequence encoding the CARHSP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11278-1mg
Specifications
CARHSP1 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CRAHSP1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol and 0.1M NaCl. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSSEPPPPPQ PPTHQASVGL LDTPRSRERS PSPLRGNVVP SPLPTRRTRT FSATVRASQG PVYKGVCKCF CRSKGHGFIT PADGGPDIFL HISDVEGEYV PVEGDEVTYK MCSIPPKNEK LQAVEVVITH LAPGTKHETW SGHVISS |